Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dmbx1
Entrez Gene ID
100770688
Gene Name
diencephalon/mesencephalon homeobox 1
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613647: 1534679 - 1540802 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003497354.2 XP_003497402.1 3 - 4 683 101
XM_003497355.1 XP_003497403.1 3 - 4 695 101

View Full Product Details