Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Shisa8
Entrez Gene ID
100763905
Gene Name
shisa family member 8
Gene Aliases
I79_021095
Chromosome Location
Chr.Un|NW_003615427: 162667 - 166419 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003512345.1 XP_003512393.1 1 - 2 55 62

View Full Product Details