Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Leap2
Entrez Gene ID
100763463
Gene Name
liver expressed antimicrobial peptide 2
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613693: 308493 - 309316 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003498505.2 XP_003498553.1 1 - 2 225 83

View Full Product Details