Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Clec3b
Entrez Gene ID
100760600
Gene Name
C-type lectin domain family 3, member B
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614144: 802936 - 809208 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003505355.2 XP_003505403.1 2 - 3 486 79

View Full Product Details