Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Prrg3
Entrez Gene ID
100772670
Gene Name
proline rich Gla (G-carboxyglutamic acid) 3 (transmembrane)
Gene Aliases
I79_018287
Chromosome Location
Chr.Un|NW_003614816: 66517 - 150135 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003510039.1 XP_003510087.1 6 - 7 1026 145

View Full Product Details