Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cxcl9
Entrez Gene ID
100770763
Gene Name
chemokine (C-X-C motif) ligand 9
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003615570: 71464 - 76555 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003512753.2 XP_003512801.1 2 - 3 210 66

View Full Product Details