Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Wnt8b
Entrez Gene ID
100775075
Gene Name
wingless-type MMTV integration site family, member 8B
Gene Aliases
I79_016390
Chromosome Location
Chr.Un|NW_003614544: 62020 - 81741 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003508442.2 XP_003508490.1 1 - 2 121 79

View Full Product Details