Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Prelid1
Entrez Gene ID
100764743
Gene Name
PRELI domain containing 1
Gene Aliases
I79_003903
Chromosome Location
Chr.Un|NW_003613681: 951264 - 953873 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007623658.1 XP_007621848.1 3 - 4 550 97
GenBank mRNA JI896801.1 - 1 - 1 539 97

View Full Product Details