Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Slc7a4
Entrez Gene ID
100757506
Gene Name
solute carrier family 7, member 4
Gene Aliases
I79_009525
Chromosome Location
Chr.Un|NW_003613919: 1507118 - 1509711 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003502841.1 XP_003502889.1 1 - 2 983 63

View Full Product Details