Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Igsf11
Entrez Gene ID
100751379
Gene Name
immunoglobulin superfamily, member 11
Gene Aliases
I79_015676
Chromosome Location
Chr.Un|NW_003614447: 633787 - 740329 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007648498.1 XP_007646688.1 2 - 3 216 105

View Full Product Details