Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pabpn1l
Entrez Gene ID
100765187
Gene Name
poly(A) binding protein, nuclear 1-like (cytoplasmic)
Gene Aliases
I79_000220
Chromosome Location
Chr.Un|NW_003613583: 909063 - 912089 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003495089.1 XP_003495137.1 3 - 4 467 111

View Full Product Details