Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ppm1l
Entrez Gene ID
100756738
Gene Name
protein phosphatase, Mg2+/Mn2+ dependent, 1L
Gene Aliases
I79_010019
Chromosome Location
Chr.Un|NW_003613954: 1010289 - 1268976 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003503263.2 XP_003503311.1 1 - 2 554 76

View Full Product Details