Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ubl4a
Entrez Gene ID
100761792
Gene Name
ubiquitin-like 4A
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003615411: 147690 - 150283 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007652804.1 XP_007650994.1 2 - 3 404 76

View Full Product Details