Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rbmx2
Entrez Gene ID
100773395
Gene Name
RNA binding motif protein, X-linked 2
Gene Aliases
I79_004956
Chromosome Location
Chr.Un|NW_003613712: 48930 - 55282 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007636315.1 XP_007634505.1 1 - 2 208 99

View Full Product Details