Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Apopt1
Entrez Gene ID
100750367
Gene Name
apoptogenic 1, mitochondrial
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613702: 1291937 - 1311809 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003498760.2 XP_003498808.2 3 - 4 329 80

View Full Product Details