Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc15a5
Entrez Gene ID
100750387
Gene Name
solute carrier family 15, member 5
Gene Aliases
I79_007225
Chromosome Location
Chr.Un|NW_003613820: 1214711 - 1310595 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003500909.1 XP_003500957.1 2 - 3 555 66

View Full Product Details