Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lyrm2
Entrez Gene ID
100772894
Gene Name
LYR motif containing 2
Gene Aliases
I79_000690
Chromosome Location
Chr.Un|NW_003613592: 1262081 - 1264812 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007641099.1 XP_007639289.1 2 - 3 217 86

View Full Product Details