Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ogfr
Entrez Gene ID
100765720
Gene Name
opioid growth factor receptor
Gene Aliases
I79_009867
Chromosome Location
Chr.Un|NW_003613943: 518011 - 524230 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007643787.1 XP_007641977.1 2 - 3 438 105
GenBank mRNA JI896721.1 - 2 - 3 250 105

View Full Product Details