Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ftl
Entrez Gene ID
100753846
Gene Name
ferritin, light polypeptide
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614570: 288958 - 290256 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007649272.1 XP_007647462.1 2 - 2 442 82
XM_007649273.1 XP_007647463.1 3 - 3 616 82

View Full Product Details