Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PDE6G
Entrez Gene ID
102171463
Gene Name
phosphodiesterase 6G
Gene Aliases
-
Chromosome Location
Chr.19: 50526546 - 50528328 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_013972601.2 XP_013828055.1 1 - 2 236 72

View Full Product Details