Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LLPH
Entrez Gene ID
102178884
Gene Name
LLP homolog, long-term synaptic facilitation factor
Gene Aliases
-
Chromosome Location
Chr.5: 46998786 - 47004955 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005680215.2 XP_005680272.1 2 - 3 472 87
GenBank mRNA JO580295.1 - 1 - 2 393 87
KA308927.1 - 1 - 2 319 87

View Full Product Details