Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C1H21orf91
Entrez Gene ID
102175802
Gene Name
chromosome 1 C21orf91 homolog
Gene Aliases
-
Chromosome Location
Chr.1: 17836540 - 17868813 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005674758.3 XP_005674815.2 3 - 4 866 64
XM_018051751.1 XP_017907240.1 3 - 4 761 64
GenBank mRNA JO329956.2 - 4 - 5 756 64
JO329958.2 - 4 - 5 756 64
JO355659.2 - 1 - 2 399 64
JO403437.2 - 2 - 3 389 64
JO455640.2 - 1 - 2 318 64
JO532561.2 - 3 - 4 741 64
JO611771.2 - 2 - 3 613 64
JO611772.2 - 2 - 3 613 64
JT738511.1 - 5 - 6 674 64
KA341029.1 - 3 - 4 736 64

View Full Product Details