Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SHOC2
Entrez Gene ID
102174087
Gene Name
SHOC2 leucine rich repeat scaffold protein
Gene Aliases
-
Chromosome Location
Chr.26: 20453264 - 20538383 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005698466.3 XP_005698523.1 8 - 9 1860 111
XM_005698467.3 XP_005698524.1 8 - 9 1851 111
XM_018041275.1 XP_017896764.1 8 - 9 1846 111
XM_018041276.1 XP_017896765.1 8 - 9 1949 111

View Full Product Details