Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KCTD8
Entrez Gene ID
102181851
Gene Name
potassium channel tetramerization domain containing 8
Gene Aliases
-
Chromosome Location
Chr.6: 63404154 - 63685945 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005681564.3 XP_005681621.2 1 - 2 1335 72

View Full Product Details