Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HYPK
Entrez Gene ID
102185509
Gene Name
huntingtin interacting protein K
Gene Aliases
-
Chromosome Location
Chr.21: 54928971 - 54930332 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005695312.3 XP_005695369.1 1 - 2 285 90
GenBank mRNA KA330286.1 - 1 - 2 220 90

View Full Product Details