Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SAMD10
Entrez Gene ID
102189598
Gene Name
sterile alpha motif domain containing 10
Gene Aliases
-
Chromosome Location
Chr.13: 53193877 - 53198629 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018057490.1 XP_017912979.1 3 - 4 750 80
XM_018057491.1 XP_017912980.1 3 - 4 750 80

View Full Product Details