Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
YPEL3
Entrez Gene ID
102186451
Gene Name
yippee like 3
Gene Aliases
-
Chromosome Location
Chr.25: 26103312 - 26106693 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA JO341697.2 - 2 - 3 799 105

View Full Product Details