Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
OAZ3
Entrez Gene ID
102191290
Gene Name
ornithine decarboxylase antizyme 3
Gene Aliases
-
Chromosome Location
Chr.3: 101142720 - 101149439 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001290529.1 NP_001277458.1 3 - 4 509 75
NM_001301483.1 NP_001288412.1 3 - 4 541 75

View Full Product Details