Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CSN3
Entrez Gene ID
100861231
Gene Name
casein kappa
Gene Aliases
CSN, CSN10, CSNK, Kappa-CN, k-casein
Chromosome Location
Chr.6: 86197262 - 86211375 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001285587.1 NP_001272516.1 4 - 5 665 110
GenBank mRNA EF564258.1 - 1 - 2 110
X60763.1 - 4 - 5 665 110

View Full Product Details