Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MAP1LC3C
Entrez Gene ID
102175424
Gene Name
microtubule associated protein 1 light chain 3 gamma
Gene Aliases
-
Chromosome Location
Chr.16: 33461573 - 33467258 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005690568.2 XP_005690625.1 2 - 3 198 111

View Full Product Details