Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PEAK1
Entrez Gene ID
102188259
Gene Name
pseudopodium enriched atypical kinase 1
Gene Aliases
-
Chromosome Location
Chr.21: 31809584 - 32133884 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018066251.1 XP_017921740.1 6 - 7 3855 73
XM_018066252.1 XP_017921741.1 5 - 6 3769 73
GenBank mRNA JT683274.1 - 4 - 5 2999 73

View Full Product Details