Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PDLIM1
Entrez Gene ID
102172031
Gene Name
PDZ and LIM domain 1
Gene Aliases
-
Chromosome Location
Chr.26: 34804078 - 34840877 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA JO611392.2 - 6 - 7 962 61

View Full Product Details