Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MIS18A
Entrez Gene ID
102177562
Gene Name
MIS18 kinetochore protein A
Gene Aliases
-
Chromosome Location
Chr.1: 1679178 - 1690534 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA KA313145.1 - 3 - 4 218 76

View Full Product Details