Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CFAP300
Entrez Gene ID
102175044
Gene Name
cilia and flagella associated protein 300
Gene Aliases
C15H11orf70
Chromosome Location
Chr.15: 75756427 - 75785107 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005689373.3 XP_005689430.2 5 - 6 633 164

View Full Product Details