Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ESM1
Entrez Gene ID
102168666
Gene Name
endothelial cell specific molecule 1
Gene Aliases
-
Chromosome Location
Chr.20: 24161661 - 24169492 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005694710.3 XP_005694767.1 2 - 3 732 84
GenBank mRNA KA321957.1 - 2 - 3 624 84

View Full Product Details