Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PEF1
Entrez Gene ID
102168870
Gene Name
penta-EF-hand domain containing 1
Gene Aliases
-
Chromosome Location
Chr.2: 13863491 - 13881226 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005676724.3 XP_005676781.2 3 - 4 562 76

View Full Product Details