Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCZ1
Entrez Gene ID
102174266
Gene Name
CCZ1 homolog, vacuolar protein trafficking and biogenesis associated
Gene Aliases
-
Chromosome Location
Chr.25: 38828497 - 38844216 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018040766.1 XP_017896255.1 10 - 11 1045 72
XM_018040767.1 XP_017896256.1 10 - 11 1131 72

View Full Product Details