Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CCDC27
Entrez Gene ID
102182524
Gene Name
coiled-coil domain containing 27
Gene Aliases
-
Chromosome Location
Chr.16: 47858257 - 47874279 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005690840.2 XP_005690897.2 1 - 2 184 54

View Full Product Details