Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AQP10
Entrez Gene ID
102180360
Gene Name
aquaporin 10
Gene Aliases
-
Chromosome Location
Chr.3: 103902046 - 103904660 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005677552.2 XP_005677609.2 3 - 4 397 84

View Full Product Details