Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LACC1
Entrez Gene ID
102177901
Gene Name
laccase domain containing 1
Gene Aliases
-
Chromosome Location
Chr.12: 72454631 - 72468980 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018056777.1 XP_017912266.1 2 - 3 689 85
XM_018056778.1 XP_017912267.1 2 - 3 1049 85

View Full Product Details