Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCL1
Entrez Gene ID
102174690
Gene Name
C-C motif chemokine ligand 1
Gene Aliases
-
Chromosome Location
Chr.19: 15422686 - 15425944 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005693213.3 XP_005693270.1 2 - 3 252 66

View Full Product Details