Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
NCCRP1
Entrez Gene ID
102182338
Gene Name
non-specific cytotoxic cell receptor protein 1 homolog (zebrafish)
Gene Aliases
-
Chromosome Location
Chr.18: 49800972 - 49804829 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018062383.1 XP_017917872.1 2 - 3 421 73
GenBank mRNA KA322420.1 - 4 - 5 1328 73

View Full Product Details