Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SH2D7
Entrez Gene ID
102187170
Gene Name
SH2 domain containing 7
Gene Aliases
-
Chromosome Location
Chr.21: 30077485 - 30088011 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005695184.2 XP_005695241.2 2 - 3 274 70

View Full Product Details