Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HRH2
Entrez Gene ID
102168263
Gene Name
histamine receptor H2
Gene Aliases
-
Chromosome Location
Chr.10: 95729963 - 95779314 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018053706.1 XP_017909195.1 2 - 3 1788 100

View Full Product Details