Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
METTL21C
Entrez Gene ID
102180054
Gene Name
methyltransferase like 21C
Gene Aliases
-
Chromosome Location
Chr.12: 7354583 - 7363816 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005687731.3 XP_005687788.2 1 - 2 648 64

View Full Product Details