Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FAU
Entrez Gene ID
102189283
Gene Name
FAU ubiquitin like and ribosomal protein S30 fusion
Gene Aliases
-
Chromosome Location
Chr.29: 43751627 - 43753326 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005699885.3 XP_005699942.1 4 - 5 448 71

View Full Product Details