Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PHOX2B
Entrez Gene ID
102176112
Gene Name
paired like homeobox 2B
Gene Aliases
-
Chromosome Location
Chr.6: 61216689 - 61220344 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005681688.3 XP_005681745.2 1 - 2 602 91

View Full Product Details