Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TPPP2
Entrez Gene ID
102170977
Gene Name
tubulin polymerization promoting protein family member 2
Gene Aliases
-
Chromosome Location
Chr.10: 76843520 - 76851134 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018054528.1 XP_017910017.1 2 - 3 388 67

View Full Product Details