Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RAB17
Entrez Gene ID
102175645
Gene Name
RAB17, member RAS oncogene family
Gene Aliases
-
Chromosome Location
Chr.3: 3694960 - 3718717 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_018040662.1 XP_017896151.1 4 - 5 810 63
XM_018040668.1 XP_017896157.1 4 - 5 722 63
GenBank mRNA JO402301.2 - 2 - 3 886 63
JO435706.2 - 4 - 5 547 63

View Full Product Details