Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FCRL4
Entrez Gene ID
102170192
Gene Name
Fc receptor like 4
Gene Aliases
-
Chromosome Location
Chr.3: 107264513 - 107288860 on Build ARS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_013976239.2 XP_013831693.2 3 - 4 614 116
GenBank mRNA JO583980.1 - 1 - 2 246 116
JT686153.1 - 4 - 5 597 116

View Full Product Details